missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chloro-5-fluorobenzaldehyde, 97%, ACROS Organics™

Chemical Identifiers

CAS 90390-49-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H4ClFO
Linear Formula FC6H3(Cl)CHO
Formula Weight 158.56
MDL Number MFCD02261734
Chemical Name or Material 3-chloro-5-fluorobenzaldehyde
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC374750200 SDS Thermo Fisher Scientific
AC374750200
Glass bottle 20g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC374750050 SDS Fisher Scientific Acros
AC374750050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

90390-49-1
100.0
C7H4ClFO
158.56
3-chloro-5-fluorobenzaldehyde
96.0
97%
FC6H3(Cl)CHO
MFCD02261734
≥96.0%

Specifications

158.56
20g
100.0
Glass bottle
Authentic
90390-49-1
MFCD02261734
≥96.0%
Low Melting Solid or Liquid
White to yellow or clear
96.0
FC6H3(Cl)CHO
C7H4ClFO
97%
3-chloro-5-fluorobenzaldehyde

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.