Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-fluorobenzaldehyde, 97%

$103.07 - $578.59

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-fluorobenzaldehyde
CAS 90390-49-1
Molecular Formula C7H4ClFO
Formula Weight 158.56
MDL Number MFCD02261734
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2648503

  SDS
alfa aesar™
H2648503
1g Each for $103.07

AAH2648509

  SDS
alfa aesar™
H2648509
10g Each for $578.59
Description & Specifications

Specifications

Formula Weight 158.56
Quantity 1g
Molecular Formula C7H4ClFO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD02261734
CAS 90390-49-1
Assay 97%
Health Hazard 1 Warning
Chemical Name or Material 3-Chloro-5-fluorobenzaldehyde
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.