Learn More
3-chloro-5-fluoroaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 374740050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-chloro-5-fluoroaniline | |
Orange to Yellow | |
100.0 | |
5 g | |
Glass bottle | |
1.5610 to 1.563 | |
1.45 | |
ClC6H3(F)NH2 | |
pubchem2953, acmc-209kdj, 3-chloro-5-fluorobenzenamine, 5-chloro-3-fluorophenylamine, ksc493m9f, 3-chloro-5-fluoro-phenylamine, buttpark 44\01-96 | |
LPIFAHAICWJMRR-UHFFFAOYSA-N | |
3-chloro-5-fluoroaniline | |
2734838 | |
97% |
4863-91-6 | |
96.0 | |
1.4500g/mL | |
Authentic | |
Liquid | |
96% min. (GC) | |
C6H5ClFN | |
MFCD03407962 | |
Solubility in water: soluble | |
C1=C(C=C(C=C1F)Cl)N | |
145.56 | |
145.56 |
Chemical Identifiers
4863-91-6 | |
145.56 | |
LPIFAHAICWJMRR-UHFFFAOYSA-N | |
2734838 | |
C1=C(C=C(C=C1F)Cl)N |
C6H5ClFN | |
MFCD03407962 | |
pubchem2953, acmc-209kdj, 3-chloro-5-fluorobenzenamine, 5-chloro-3-fluorophenylamine, ksc493m9f, 3-chloro-5-fluoro-phenylamine, buttpark 44\01-96 | |
3-chloro-5-fluoroaniline |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
Recommended Storage : Darkens on exposure to light and air
RUO – Research Use Only