missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-5'-fluoroacetophenone, 97%

$109.34 - $993.30

Chemical Identifiers

CAS 842140-52-7
Molecular Formula C8H6ClFO
Formula Weight 172.59
MDL Number MFCD04115834
Chemical Name or Material 3'-Chloro-5'-fluoroacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2680803 SDS Alfa Aesar™
H2680803
1g Each for $109.34
AAH2680806 SDS Alfa Aesar™
H2680806
5g Each for $326.11
AAH2680814 SDS Alfa Aesar™
H2680814
25g Each for $993.30
Specifications

Description

Solubility
Soluble in diethyl ether

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

842140-52-7
172.59
3'-Chloro-5'-fluoroacetophenone
C8H6ClFO
MFCD04115834

Specifications

51° to 54°C
842140-52-7
172.59
3'-Chloro-5'-fluoroacetophenone
1g
C8H6ClFO
MFCD04115834
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning