Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-fluoro-4-methoxyphenol, 97%

$299.85 - $299.85

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-fluoro-4-methoxyphenol
Molecular Formula C7H6ClFO2
Formula Weight 176.57
MDL Number MFCD09832264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3180103

  SDS
alfa aesar™
H3180103
1g Each for $299.85
Description & Specifications

Specifications

Formula Weight 176.57
MDL Number MFCD09832264
Chemical Name or Material 3-Chloro-5-fluoro-4-methoxyphenol
Health Hazard 1 Warning
Quantity 1g
Assay 97%
Molecular Formula C7H6ClFO2
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 62° to 65°C
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.