Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-fluoro-4-methoxybenzoic acid, 97%

$124.68 - $374.04

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-fluoro-4-methoxybenzoic acid
CAS 886497-22-9
Molecular Formula C8H6ClFO3
Formula Weight 204.59
MDL Number MFCD04115864
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2639703

 
alfa aesar™
H2639703
1g Each for $124.68

AAH2639706

 
alfa aesar™
H2639706
5g Each for $374.04
Description & Specifications

Specifications

Formula Weight 204.59
Chemical Name or Material 3-Chloro-5-fluoro-4-methoxybenzoic acid
Assay 97%
Molecular Formula C8H6ClFO3
MDL Number MFCD04115864
CAS 886497-22-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.