Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-fluoro-4-methoxybenzaldehyde, 97%

$189.85 - $578.59

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-fluoro-4-methoxybenzaldehyde
CAS 177994-49-9
Molecular Formula C8H6ClFO2
Formula Weight 188.59
MDL Number MFCD04115863
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2666603

  SDS
alfa aesar™
H2666603
1g Each for $189.85

AAH2666606

  SDS
alfa aesar™
H2666606
5g Each for $578.59
Description & Specifications

Specifications

Formula Weight 188.59
Assay 97%
Melting Point 43° to 45°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 177994-49-9
Chemical Name or Material 3-Chloro-5-fluoro-4-methoxybenzaldehyde
Molecular Formula C8H6ClFO2
MDL Number MFCD04115863
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.