Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-5'-fluoro-4'-methoxyacetophenone, 97%

$136.58 - $136.58

Chemical Identifiers

Chemical Name or Material 3'-Chloro-5'-fluoro-4'-methoxyacetophenone
CAS 886497-19-4
Molecular Formula C9H8ClFO2
Formula Weight 202.61g/mol
MDL Number MFCD04115862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2660206

 
alfa aesar™
H26602-06
5g Each for $136.58
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 3'-Chloro-5'-fluoro-4'-methoxyacetophenone
CAS 886497-19-4
Assay 97%
Molecular Formula C9H8ClFO2
Formula Weight 202.61g/mol
MDL Number MFCD04115862
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.