Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5,5-dimethyl-2-cyclohexen-1-one, 96%

$70.35 - $70.35

Chemical Identifiers

Chemical Name or Material 3-Chloro-5,5-dimethyl-2-cyclohexen-1-one
CAS 17530-69-7
Molecular Formula C8H11ClO
Formula Weight 158.63
MDL Number MFCD00051620
Beilstein 1099253
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1072806

  SDS
alfa aesar™
A1072806
5g Each for $70.35
Description & Specifications

Specifications

Formula Weight 158.63
Chemical Name or Material 3-Chloro-5,5-dimethyl-2-cyclohexen-1-one
Assay 96%
Molecular Formula C8H11ClO
MDL Number MFCD00051620
CAS 17530-69-7
Beilstein 1099253
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2°C to 8°C
Boiling Point 44° to 46°C (0.1mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C