missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-(trifluoromethyl)benzoyl chloride, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar H3234303

 View more versions of this product

Catalog No. AAH3234303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Chloro-4-(trifluoromethyl)benzoyl chloride
Colorless
1092460-77-9
97%
MFCD11519325
KKXPAIGCYQCQHM-UHFFFAOYSA-N
3-chloro-4-(trifluoromethyl)benzoyl chloride
21275239
Liquid
Moisture Sensitive
1g
1.5030
C8H3Cl2F3O
UN3265
C1=CC(=C(C=C1C(=O)Cl)Cl)C(F)(F)F
243.006
243.01

Chemical Identifiers

1092460-77-9
243.006
KKXPAIGCYQCQHM-UHFFFAOYSA-N
C8H3Cl2F3O
MFCD11519325
21275239
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents