Please login to your online account to display your discounted pricing

3-Chloro-4-(trifluoromethyl)benzoyl chloride, 97%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzoyl chloride
Molecular Formula C8H3Cl2F3O
Formula Weight 243.01
MDL Number MFCD11519325
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH3234303

  SDS
alfa aesar
H3234303
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3234306

  SDS
alfa aesar
H3234306
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 243.01
Quantity 1g
Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzoyl chloride
Refractive Index 1.5030
Physical Form Liquid
Assay 97%
Molecular Formula C8H3Cl2F3O
MDL Number MFCD11519325
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Sensitivity Moisture Sensitive
Color Colorless
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.