missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-(trifluoromethyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H53120MD

 View more versions of this product

Catalog No. AAH53120MD



Specifications

Specifications

3-Chloro-4-(trifluoromethyl)benzeneboronic acid
250mg
C7H5BClF3O2
3-chloro-4-trifluoromethyl phenylboronic acid, 3-chloro-4-trifluoromethylphenylboronic acid, 3-chloro-4-trifluoromethyl benzeneboronic acid, 3-chloro-4-trifluoromethyl phenyl boronic acid, 4-borono-2-chlorobenzotrifluoride, boronic acid, 3-chloro-4-trifluoromethyl phenyl, 3-chloro-4-trifluoromethyl phenyl boronic acid, 3-chloro-4-trifluoromethyl benzeneboronic acid, pubchem23707, acmc-209pwg
B(C1=CC(=C(C=C1)C(F)(F)F)Cl)(O)O
224.37
224.38
206°C to 212°C
847756-88-1
MFCD03788417
OCHKTAGGJMWISO-UHFFFAOYSA-N
[3-chloro-4-(trifluoromethyl)phenyl]boronic acid
3772958
97%

Chemical Identifiers

847756-88-1
224.37
OCHKTAGGJMWISO-UHFFFAOYSA-N
3772958
B(C1=CC(=C(C=C1)C(F)(F)F)Cl)(O)O
C7H5BClF3O2
MFCD03788417
3-chloro-4-trifluoromethyl phenylboronic acid, 3-chloro-4-trifluoromethylphenylboronic acid, 3-chloro-4-trifluoromethyl benzeneboronic acid, 3-chloro-4-trifluoromethyl phenyl boronic acid, 4-borono-2-chlorobenzotrifluoride, boronic acid, 3-chloro-4-trifluoromethyl phenyl, 3-chloro-4-trifluoromethyl phenyl boronic acid, 3-chloro-4-trifluoromethyl benzeneboronic acid, pubchem23707, acmc-209pwg
[3-chloro-4-(trifluoromethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents