Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%

$311.91 - $311.91

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzaldehyde
Molecular Formula C8H4ClF3O
Formula Weight 208.57
MDL Number MFCD03095197
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3163503

  SDS
alfa aesar™
H3163503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3163506

  SDS
alfa aesar™
H3163506
5g Each for $311.91
Description & Specifications

Specifications

Formula Weight 208.57
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzaldehyde
Molecular Formula C8H4ClF3O
MDL Number MFCD03095197
Health Hazard 1 Warning

Applications
3-Chloro-4-(trifluoromethyl)benzaldehyde is used in synthesis of organic compunds in γ-Aminobutyrate-A Receptor Modulation by 3-Aryl-1-(arylsulfonyl)- 1,4,5,6-tetrahydropyridazines.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.