missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-nitrotoluene, 98%, ACROS Organics™

$281.17 - $281.17

Chemical Identifiers

CAS 38939-88-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD02683043
InChI Key KGSQRFPDZCBVBS-UHFFFAOYSA-N
Synonym 3-chloro-4-nitrotoluene, 2-chloro-4-methyl-1-nitro-benzene, benzene, 2-chloro-4-methyl-1-nitro, pubchem4534, acmc-1afh8, ksc493s6j, 3-chloro-4-nitrotoluene
PubChem CID 123478
IUPAC Name 2-chloro-4-methyl-1-nitrobenzene
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC348080050 SDS View Documents Acros Organics
348080050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC348080250 SDS View Documents Acros Organics
348080250
25g Glass bottle Each for $281.17
Specifications

Chemical Identifiers

38939-88-7
171.58
KGSQRFPDZCBVBS-UHFFFAOYSA-N
123478
CC1=CC(=C(C=C1)[N+](=O)[O-])Cl
C7H6ClNO2
MFCD02683043
3-chloro-4-nitrotoluene, 2-chloro-4-methyl-1-nitro-benzene, benzene, 2-chloro-4-methyl-1-nitro, pubchem4534, acmc-1afh8, ksc493s6j, 3-chloro-4-nitrotoluene
2-chloro-4-methyl-1-nitrobenzene

Specifications

>230°C
Glass bottle
108.0°C (3.0 mmHg)
22.0°C
38939-88-7
100.0
C7H6ClNO2
MFCD02683043
3-chloro-4-nitrotoluene, 2-chloro-4-methyl-1-nitro-benzene, benzene, 2-chloro-4-methyl-1-nitro, pubchem4534, acmc-1afh8, ksc493s6j, 3-chloro-4-nitrotoluene
KGSQRFPDZCBVBS-UHFFFAOYSA-N
2-chloro-4-methyl-1-nitrobenzene
123478
Liquid After Melting
3-Chloro-4-nitrotoluene
Authentic
1.5610 to 1.5640
Yellow
5g
97.5
98%
CH3C6H3(NO2)Cl
05, IV, 856
Solubility in water: insoluble.
CC1=CC(=C(C=C1)[N+](=O)[O-])Cl
171.58
171.58
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 254-199-6

TSCA : TSCA