Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-nitrobenzotrifluoride, 95%

$91.30 - $274.23

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-nitrobenzotrifluoride
CAS 402-11-9
Molecular Formula C7H3ClF3NO2
Formula Weight 225.55
MDL Number MFCD00272564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2838827

  SDS
alfa aesar™
H2838827
2.5g Each for $274.23

AAH28388ME

  SDS
alfa aesar™
H28388ME
500mg Each for $91.30
Description & Specifications

Specifications

Formula Weight 225.55
Boiling Point 54°C (0.4mmHg)
Chemical Name or Material 3-Chloro-4-nitrobenzotrifluoride
Assay 95%
Quantity 2.5g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Molecular Formula C7H3ClF3NO2
CAS 402-11-9
MDL Number MFCD00272564
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.