Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-nitrobenzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-nitrobenzenesulfonyl chloride
CAS 64835-30-9
Molecular Formula C6H3Cl2NO4S
Formula Weight 256.07
MDL Number MFCD04973767
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2611503

  SDS
alfa aesar™
H2611503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH26115MD

  SDS
alfa aesar™
H26115MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.07
CAS 64835-30-9
Melting Point 36° to 39°C
Molecular Formula C6H3Cl2NO4S
UN Number UN3261
Health Hazard 1 Danger
Chemical Name or Material 3-Chloro-4-nitrobenzenesulfonyl chloride
Assay 98%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
MDL Number MFCD04973767
Quantity 1g

Applications
3-Chloro-4-nitrobenzenesulfonyl chloride is the reactant in the synthesis of aminobenzenesulfonyl fluorides. 3-Chloro-4-nitrobenzenesulfonyl chloride is also used as intermediates in organic synthesis.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.