missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylpyridine, 95%

Manufacturer:  Alfa Aesar™ H6162614

 View more versions of this product

Catalog No. AAH6162614



Specifications

Specifications

3-Chloro-4-methylpyridine
25g
C6H6ClN
3-chloro-4-picoline, pyridine, 3-chloro-4-methyl, 3-chloro-4-methyl-pyridine, pubchem14323, acmc-1bl3c, 3-chloro-4-methylpyridine?, ksc494c8t, 3-chloro-4-methylpyridine
CC1=C(C=NC=C1)Cl
127.571
127.57
95%
Colorless to Brown
72093-04-0
MFCD04114245
VLNXNJMHLZFECV-UHFFFAOYSA-N
3-chloro-4-methylpyridine
2762925
Liquid

Chemical Identifiers

72093-04-0
127.571
VLNXNJMHLZFECV-UHFFFAOYSA-N
2762925
CC1=C(C=NC=C1)Cl
C6H6ClN
MFCD04114245
3-chloro-4-picoline, pyridine, 3-chloro-4-methyl, 3-chloro-4-methyl-pyridine, pubchem14323, acmc-1bl3c, 3-chloro-4-methylpyridine?, ksc494c8t, 3-chloro-4-methylpyridine
3-chloro-4-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents