Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylphenylhydrazine hydrochloride, 97%

$50.47 - $50.47

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-methylphenylhydrazine hydrochloride
CAS 54812-56-5
Molecular Formula C7H9ClN2·HCl
Formula Weight 193.08
MDL Number MFCD00012936
Synonym 3-Chloro-p-tolylhydrazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2205303

  SDS
alfa aesar™
B2205303
1g Each for $50.47
Description & Specifications

Specifications

Formula Weight 193.08
Chemical Name or Material 3-Chloro-4-methylphenylhydrazine hydrochloride
CAS 54812-56-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Chloro-p-tolylhydrazine hydrochloride
Health Hazard 1 Warning
Molecular Formula C7H9ClN2·HCl
Assay 97%
MDL Number MFCD00012936
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈233°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.