missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylphenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1206414

 View more versions of this product

Catalog No. AAL1206414



Specifications

Specifications

3-Chloro-4-methylphenyl isocyanate
225°C
25g
C8H6ClNO
UN2236
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
CC1=C(C=C(C=C1)N=C=O)Cl
167.592
167.6
109°C (228°F)
20°C to 24°C
28479-22-3
MFCD00013858
2690771
UKTKKMZDESVUEE-UHFFFAOYSA-N
2-chloro-4-isocyanato-1-methylbenzene
62832
98%

Chemical Identifiers

28479-22-3
167.592
UKTKKMZDESVUEE-UHFFFAOYSA-N
62832
CC1=C(C=C(C=C1)N=C=O)Cl
C8H6ClNO
MFCD00013858
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
2-chloro-4-isocyanato-1-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H314-H318
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-CHLORO-4-METHYLPHENYL ISOCYANATE, LIQUID

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents