missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-methylbenzyl alcohol, tech., mixture of isomers, Thermo Scientific™

$134.74 - $528.37

Chemical Identifiers

CAS 39652-32-9
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00016863
InChI Key NLRJZTGNCBMNKS-UHFFFAOYSA-N
PubChem CID 123486
IUPAC Name (3-chloro-4-methylphenyl)methanol
SMILES CC1=CC=C(CO)C=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4381906
View Documents Promotion Details
Thermo Scientific Chemicals
4381906
5 g
Each for $134.74
 
AA4381914
View Documents Promotion Details
Thermo Scientific Chemicals
4381914
25 g
Each for $528.37
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

39652-32-9
156.61
NLRJZTGNCBMNKS-UHFFFAOYSA-N
(3-chloro-4-methylphenyl)methanol
C8H9ClO
MFCD00016863
123486
CC1=CC=C(CO)C=C1Cl

Specifications

39652-32-9
MFCD00016863
3-chloro-4-methylbenzyl alcohol, 3-chloro-4-methylphenyl methanol, benzenemethanol, 3-chloro-4-methyl, 3-chloro-4-methylphenyl methan-1-ol, 3-chloro-4-methylbenzyl pound inverted question markalcohol, 3-chloro-4-methylbenzylalcohol, pubchem23926, acmc-20an6l, benzenemethanol,3-chloro-4-methyl, 3-chloro-4-methylphenyl methanol #
CC1=CC=C(CO)C=C1Cl
156.61
156.61
Liquid
C8H9ClO
5 g
NLRJZTGNCBMNKS-UHFFFAOYSA-N
(3-chloro-4-methylphenyl)methanol
123486
Technical
3-Chloro-4-methylbenzyl alcohol, Mixture of isomers
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 254-566-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'