missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylbenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5583303

 View more versions of this product

Catalog No. AAH5583303



Specifications

Specifications

3-Chloro-4-methylbenzonitrile
45°C to 48°C
21423-81-4
MFCD00001799
INEMHABDFCKBID-UHFFFAOYSA-N
3-chloro-4-methylbenzonitrile
88894
97%
84°C (183°F)
1g
C8H6ClN
2-chloro-4-cyanotoluene
CC1=C(C=C(C=C1)C#N)Cl
151.593
151.6

Chemical Identifiers

21423-81-4
151.593
INEMHABDFCKBID-UHFFFAOYSA-N
88894
CC1=C(C=C(C=C1)C#N)Cl
C8H6ClN
MFCD00001799
2-chloro-4-cyanotoluene
3-chloro-4-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 244-381-3

TSCA : No

Recommended Storage : Ambient temperatures

Documents