Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylbenzoic acid, 97%

$62.34 - $224.73

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-methylbenzoic acid
CAS 5162-82-3
Molecular Formula C8H7ClO2
Formula Weight 170.6
MDL Number MFCD00045841
Synonym 3-Chloro-p-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2122803

 
alfa aesar™
B2122803
1g Each for $62.34

AAB2122806

 
alfa aesar™
B2122806
5g Each for $224.73
Description & Specifications

Specifications

Formula Weight 170.6
Chemical Name or Material 3-Chloro-4-methylbenzoic acid
Assay 97%
Molecular Formula C8H7ClO2
MDL Number MFCD00045841
CAS 5162-82-3
Synonym 3-Chloro-p-toluic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 198° to 200°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.