Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylbenzenesulfonyl chloride, 99%

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-methylbenzenesulfonyl chloride
CAS 42413-03-6
Molecular Formula C7H6Cl2O2S
Formula Weight 225.09
MDL Number MFCD00203987
UN Number UN3261
Beilstein 2364094
Synonym 3-Chloro-p-toluenesulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1988603

  SDS
alfa aesar™
L1988603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1988606

  SDS
alfa aesar™
L1988606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1988614

  SDS
alfa aesar™
L1988614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.09
Chemical Name or Material 3-Chloro-4-methylbenzenesulfonyl chloride
Assay 99%
Molecular Formula C7H6Cl2O2S
MDL Number MFCD00203987
CAS 42413-03-6
Synonym 3-Chloro-p-toluenesulfonyl chloride
Health Hazard 1 Danger
UN Number UN3261
Beilstein 2364094
Health Hazard 2 GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P280-P273-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 155° to 156°C (19mmHg)
Melting Point 34° to 38°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P280-P273-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.