missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylanisole, 97%

$85.88 - $275.24

Chemical Identifiers

CAS 54788-38-4
Molecular Formula C8H9ClO
Formula Weight 156.61
MDL Number MFCD00087376
Synonym 2-Chloro-4-methoxytoluene
Chemical Name or Material 3-Chloro-4-methylanisole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2740106 SDS Alfa Aesar™
H2740106
5g Each for $85.88
AAH2740114 SDS Alfa Aesar™
H2740114
25g Each for $275.24
Specifications Description & Specifications

Chemical Identifiers

54788-38-4
156.61
2-Chloro-4-methoxytoluene
C8H9ClO
MFCD00087376
3-Chloro-4-methylanisole

Specifications

76°C (2mmHg)
54788-38-4
156.61
2-Chloro-4-methoxytoluene
97%
5g
C8H9ClO
MFCD00087376
3-Chloro-4-methylanisole

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger