missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 3-Chloro-4-methylaniline hydrochloride, 97%
$44.47 - $397.09
Chemical Identifiers
CAS | 7745-89-3 |
---|---|
Molecular Formula | C7H9Cl2N |
Molecular Weight (g/mol) | 178.056 |
MDL Number | MFCD00242925 |
InChI Key | YOQUZCNSUOJWQS-UHFFFAOYSA-N |
Synonym | 3-chloro-4-methylaniline hydrochloride, 3-chloro-p-toluidine hydrochloride, 4-methyl, 3-chloroaniline hydrochloride, benzenamine, 3-chloro-4-methyl-, hydrochloride, caswell no. 216a, cth, epa pesticide chemical code 009901, p-toluidine, 3-chloro-, hydrochloride, starlicide, unii-7qq8474d5s |
PubChem CID | 62651 |
IUPAC Name | 3-chloro-4-methylaniline;hydrochloride |
SMILES | CC1=C(C=C(C=C1)N)Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3134206
|
Alfa Aesar™
H3134206 |
5g |
Each for $44.47
|
|
AAH3134214
|
Alfa Aesar™
H3134214 |
25g |
Each for $142.64
|
|
AAH3134222
|
Alfa Aesar™
H3134222 |
100g |
Each for $397.09
|
|
Chemical Identifiers
7745-89-3 | |
178.056 | |
YOQUZCNSUOJWQS-UHFFFAOYSA-N | |
62651 | |
CC1=C(C=C(C=C1)N)Cl.Cl |
C7H9Cl2N | |
MFCD00242925 | |
3-chloro-4-methylaniline hydrochloride, 3-chloro-p-toluidine hydrochloride, 4-methyl, 3-chloroaniline hydrochloride, benzenamine, 3-chloro-4-methyl-, hydrochloride, caswell no. 216a, cth, epa pesticide chemical code 009901, p-toluidine, 3-chloro-, hydrochloride, starlicide, unii-7qq8474d5s | |
3-chloro-4-methylaniline;hydrochloride |
Specifications
~260°C (sublimation) | |
7745-89-3 | |
MFCD00242925 | |
YOQUZCNSUOJWQS-UHFFFAOYSA-N | |
3-chloro-4-methylaniline;hydrochloride | |
62651 | |
97% |
5g | |
C7H9Cl2N | |
3-chloro-4-methylaniline hydrochloride, 3-chloro-p-toluidine hydrochloride, 4-methyl, 3-chloroaniline hydrochloride, benzenamine, 3-chloro-4-methyl-, hydrochloride, caswell no. 216a, cth, epa pesticide chemical code 009901, p-toluidine, 3-chloro-, hydrochloride, starlicide, unii-7qq8474d5s | |
CC1=C(C=C(C=C1)N)Cl.Cl | |
178.056 | |
178.06 | |
3-Chloro-4-methylaniline hydrochloride |
Safety and Handling
GHS H Statement
H373-H302-H312-H332-H315-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H373
EINECSNumber : 202-446-3
RTECSNumber : XU5425000
TSCA : No
Recommended Storage : Ambient temperatures