Please login to your online account to display your discounted pricing

3-Chloro-4-methylaniline 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  146502500

 View more versions of this product

Catalog No. AC146502500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 141.6
Quantity 250g
EINECS Number 202-446-3
TSCA TSCA
RTECS Number XU5111000
Beilstein 12, 988
CAS Max % 100.0
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Packaging Glass bottle
Chemical Name or Material 3-Chloro-4-methylaniline
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
CAS 95-74-9
CAS Min % 98.5
Name Note 99%
Assay Percent Range 99%
Synonym 3-Chloro-p-toluidine
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Chloro-4-methylaniline
Name Note 99%
CAS 95-74-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Formula Weight 141.6
Synonym 3-Chloro-p-toluidine
Accessories

GHS Signal Word: Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents