Please login to your online account to display your discounted pricing

3-Chloro-4-methylaniline 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-methylaniline
Name Note 99%
CAS 95-74-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Formula Weight 141.6
Synonym 3-Chloro-p-toluidine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146500010

  SDS
thermo fisher scientific
146500010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC146502500

  SDS
thermo fisher scientific
146502500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.6
Quantity 1kg
Beilstein 12, 988
TSCA TSCA
EINECS Number 202-446-3
RTECS Number XU5111000
Density 1.1700g/mL
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Chloro-4-methylaniline
Name Note 99%
Linear Formula ClC6H3(CH3)NH2
CAS Min % 98.5
CAS 95-74-9
Assay Percent Range 99%
Molecular Formula C7H8ClN
Synonym 3-Chloro-p-toluidine
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.