missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methylaniline, 98%

Manufacturer:  Alfa Aesar™ A154390I

 View more versions of this product

Catalog No. AAA154390I



Specifications

Specifications

3-Chloro-4-methylaniline
236°C to 238°C
5000g
C7H8ClN
UN2239
3-chloro-p-toluidine, 2-chloro-4-aminotoluene, 4-amino-2-chlorotoluene, benzenamine, 3-chloro-4-methyl, 3-chloro-4-methylbenzenamine, 3-chloro-4-methylphenylamine, 1-amino-3-chloro-4-methylbenzene, 4-methyl-3-chloroaniline, p-toluidine, 3-chloro, gull toxicant
CC1=C(C=C(C=C1)N)Cl
141.598
CHEBI:37824
98%
130°C (266°F)
22°C to 24°C
95-74-9
MFCD00007773
636511
RQKFYFNZSHWXAW-UHFFFAOYSA-N
3-chloro-4-methylaniline
7255
141.6

Chemical Identifiers

95-74-9
141.598
RQKFYFNZSHWXAW-UHFFFAOYSA-N
7255
3-chloro-4-methylaniline
C7H8ClN
MFCD00007773
3-chloro-p-toluidine, 2-chloro-4-aminotoluene, 4-amino-2-chlorotoluene, benzenamine, 3-chloro-4-methyl, 3-chloro-4-methylbenzenamine, 3-chloro-4-methylphenylamine, 1-amino-3-chloro-4-methylbenzene, 4-methyl-3-chloroaniline, p-toluidine, 3-chloro, gull toxicant
CHEBI:37824
CC1=C(C=C(C=C1)N)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID

RTECSNumber : XU5111000

TSCA : TSCA

Documents