Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-4'-methylacetanilide, 98%

$44.15 - $159.72

Chemical Identifiers

Chemical Name or Material 3'-Chloro-4'-methylacetanilide
CAS 7149-79-3
Molecular Formula C9H10ClNO
Formula Weight 183.64
MDL Number MFCD00032582
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1933614

  SDS
alfa aesar™
A1933614
25g Each for $159.72

AAA1933606

  SDS
alfa aesar™
A1933606
5g Each for $44.15
Description & Specifications

Specifications

Formula Weight 183.64
Chemical Name or Material 3'-Chloro-4'-methylacetanilide
TSCA TSCA
Assay 98%
Molecular Formula C9H10ClNO
MDL Number MFCD00032582
CAS 7149-79-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AN3325000
Melting Point 102° to 105°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.