Please login to your online account to display your discounted pricing

3-chloro-4-methyl-7-hydroxycoumarin, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-chloro-4-methyl-7-hydroxycoumarin
CAS 6174-86-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H7ClO3
Formula Weight 210.62
MDL Number MFCD00006851
Synonym 3-Chloro-4-methylumbelliferone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC345340050

  SDS
thermo fisher scientific
AC345340050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC345340010

  SDS
thermo fisher scientific
AC345340010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 210.62
Quantity 5g
Color White to off-white
Melting Point 244° to 248°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-chloro-4-methyl-7-hydroxycoumarin
Molecular Formula C10H7ClO3
MDL Number MFCD00006851
CAS 6174-86-3
Synonym 3-Chloro-4-methylumbelliferone
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass bottle