Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methoxypyridine, 98%

$88.59 - $88.59

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-methoxypyridine
CAS 89379-79-3
Molecular Formula C6H6ClNO
Formula Weight 143.57
MDL Number MFCD09743650
Beilstein 116431
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH6429803

  SDS
alfa aesar™
H6429803
Each for $88.59

AAH6429806

  SDS
alfa aesar™
H6429806
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64298MD

  SDS
alfa aesar™
H64298MD
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.57
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Chloro-4-methoxypyridine
Assay 98%
Molecular Formula C6H6ClNO
CAS 89379-79-3
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD09743650
Recommended Storage 2° to 8°C
Beilstein 116431
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C