missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chloro-4-methoxyphenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 175883-60-0
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C7H8BClO3
Linear Formula CH3OC6H3(Cl)B(OH)2
Formula Weight 186.4
MDL Number MFCD04039888
Chemical Name or Material 3-Chloro-4-methoxyphenylboronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC427920010 Thermo Fisher Scientific
AC427920010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427920050 Thermo Fisher Scientific
AC427920050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

175883-60-0
100.0
C7H8BClO3
186.4
3-Chloro-4-methoxyphenylboronic acid
94.0
95%
CH3OC6H3(Cl)B(OH)2
MFCD04039888

Specifications

1g
94.0
Glass bottle
C7H8BClO3
186.4
3-Chloro-4-methoxyphenylboronic acid
175883-60-0
100.0
95%
CH3OC6H3(Cl)B(OH)2
MFCD04039888

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.