missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloro-4-methoxybenzylamine Hydrochloride 97.0+%, TCI America™
$28.21 - $81.29
Chemical Identifiers
CAS | 41965-95-1 |
---|---|
Molecular Formula | C8H11Cl2NO |
Molecular Weight (g/mol) | 208.082 |
MDL Number | MFCD01211460 |
InChI Key | IKWWOZCEHOYKAO-UHFFFAOYSA-N |
Synonym | 3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride |
PubChem CID | 2764287 |
IUPAC Name | (3-chloro-4-methoxyphenyl)methanamine;hydrochloride |
SMILES | COC1=C(C=C(C=C1)CN)Cl.Cl |
Chemical Identifiers
41965-95-1 | |
208.082 | |
IKWWOZCEHOYKAO-UHFFFAOYSA-N | |
2764287 | |
COC1=C(C=C(C=C1)CN)Cl.Cl |
C8H11Cl2NO | |
MFCD01211460 | |
3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride | |
(3-chloro-4-methoxyphenyl)methanamine;hydrochloride |
Specifications
41965-95-1 | |
White | |
MFCD01211460 | |
3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride | |
COC1=C(C=C(C=C1)CN)Cl.Cl | |
208.082 | |
208.08 | |
Crystalline Powder |
252°C | |
C8H11Cl2NO | |
1 g | |
IKWWOZCEHOYKAO-UHFFFAOYSA-N | |
(3-chloro-4-methoxyphenyl)methanamine;hydrochloride | |
2764287 | |
≥97.0% (HPLC,T) | |
3-Chloro-4-methoxybenzylamine Hydrochloride |
Safety and Handling
TSCA : No