missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methoxybenzylamine hydrochloride, 97%

$38.41 - $564.42

Chemical Identifiers

CAS 41965-95-1
Molecular Formula C8H11Cl2NO
Molecular Weight (g/mol) 208.082
MDL Number MFCD01211460
InChI Key IKWWOZCEHOYKAO-UHFFFAOYSA-N
Synonym 3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride
PubChem CID 2764287
IUPAC Name (3-chloro-4-methoxyphenyl)methanamine;hydrochloride
SMILES COC1=C(C=C(C=C1)CN)Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6380603 View Documents Alfa Aesar™
H6380603
1g Each for $38.41
 
AAH6380606 View Documents Alfa Aesar™
H6380606
5g Each for $141.88
 
AAH6380614 View Documents Alfa Aesar™
H6380614
25g Each for $564.42
 
Specifications

Chemical Identifiers

41965-95-1
208.082
IKWWOZCEHOYKAO-UHFFFAOYSA-N
2764287
COC1=C(C=C(C=C1)CN)Cl.Cl
C8H11Cl2NO
MFCD01211460
3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride
(3-chloro-4-methoxyphenyl)methanamine;hydrochloride

Specifications

250°C to 255°C
41965-95-1
MFCD01211460
3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride
COC1=C(C=C(C=C1)CN)Cl.Cl
208.082
208.09
97%
1g
C8H11Cl2NO
UN2811
IKWWOZCEHOYKAO-UHFFFAOYSA-N
(3-chloro-4-methoxyphenyl)methanamine;hydrochloride
2764287
Solid
3-Chloro-4-methoxybenzylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.