missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-methoxybenzylamine hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6380603

 View more versions of this product

Catalog No. AAH6380603



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Chloro-4-methoxybenzylamine hydrochloride
250°C to 255°C
MFCD01211460
UN2811
IKWWOZCEHOYKAO-UHFFFAOYSA-N
(3-chloro-4-methoxyphenyl)methanamine;hydrochloride
2764287
97%
41965-95-1
C8H11Cl2NO
1 g
3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride
COC1=C(C=C(C=C1)CN)Cl.Cl
208.082
208.09
Solid

Chemical Identifiers

41965-95-1
208.082
IKWWOZCEHOYKAO-UHFFFAOYSA-N
2764287
COC1=C(C=C(C=C1)CN)Cl.Cl
C8H11Cl2NO
MFCD01211460
3-chloro-4-methoxybenzylamine hydrochloride, 3-chloro-4-methoxyphenyl methanamine hydrochloride, 3-chloro-4-methoxybenzylamine hcl, 4-aminomethyl-2-chloroanisole hcl, 4-aminomethyl-2-chloroanisole hydrochloride, 1-3-chloro-4-methoxyphenyl methanamine hydrochloride, acmc-1ambw, 3-chloro-4-methoxybenzylamine.hcl, benzenemethanamine, 3-chloro-4-methoxy-, hydrochloride 1:1, chloromethoxybenzylaminehydrochloride
(3-chloro-4-methoxyphenyl)methanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H301-H315-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions