missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methoxybenzonitrile, 97+%

$26.53 - $90.68

Chemical Identifiers

CAS 102151-33-7
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD03093073
InChI Key SUFOLDHSHRVSQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-chloro-4-methoxy, 3-chloro-4-methoxybenzenecarbonitrile, pubchem3661, zlchem 1137, acmc-20a6ct, ksc494m4t, 3-chloro-4-methoxy-benzonitrile, 3-chloro-4-methoxybenzonitrile, 3-chloranyl-4-methoxy-benzenecarbonitrile
PubChem CID 4323105
IUPAC Name 3-chloro-4-methoxybenzonitrile
SMILES COC1=C(C=C(C=C1)C#N)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1850903 View Documents Alfa Aesar™
L1850903
1g Each for $26.53
 
AAL1850906 View Documents Alfa Aesar™
L1850906
5g Each for $90.68
 
Specifications

Chemical Identifiers

102151-33-7
167.592
SUFOLDHSHRVSQV-UHFFFAOYSA-N
4323105
COC1=C(C=C(C=C1)C#N)Cl
C8H6ClNO
MFCD03093073
benzonitrile, 3-chloro-4-methoxy, 3-chloro-4-methoxybenzenecarbonitrile, pubchem3661, zlchem 1137, acmc-20a6ct, ksc494m4t, 3-chloro-4-methoxy-benzonitrile, 3-chloro-4-methoxybenzonitrile, 3-chloranyl-4-methoxy-benzenecarbonitrile
3-chloro-4-methoxybenzonitrile

Specifications

108°C to 109°C
102151-33-7
MFCD03093073
5926726
SUFOLDHSHRVSQV-UHFFFAOYSA-N
3-chloro-4-methoxybenzonitrile
4323105
≥97%
1g
C8H6ClNO
UN3439
benzonitrile, 3-chloro-4-methoxy, 3-chloro-4-methoxybenzenecarbonitrile, pubchem3661, zlchem 1137, acmc-20a6ct, ksc494m4t, 3-chloro-4-methoxy-benzonitrile, 3-chloro-4-methoxybenzonitrile, 3-chloranyl-4-methoxy-benzenecarbonitrile
COC1=C(C=C(C=C1)C#N)Cl
167.592
167.6
3-Chloro-4-methoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.