missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chloro-4-methoxybenzaldehyde, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC435310050

 View more versions of this product

Catalog No. AC435310050



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-chloro-4-methoxybenzaldehyde
96.0
Brown to White
128°C (5.0mmHg)
Authentic
MFCD00016601
2-ethyl pyromeconic acid, 2-ethyl-3-hydroxy-4-pyrone, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethylpyromeconic acid, 3-hydroxy-2-ethyl-1,4-pyrone, 3-hydroxy-2-ethyl-4-pyrone, 4h-pyran-4-one, 2-ethyl-3-hydroxy, ethyl maltol, ethylmaltol, veltol plus
WYVGYYIZXPXHAZ-UHFFFAOYSA-N
3-chloro-4-methoxybenzaldehyde
21059
Powder
4903-09-7
100.0
54°C to 56°C
96% min. (GC)
C8H7ClO2
Glass bottle
5g
COC1=CC=C(C=O)C=C1Cl
170.6
170.6
97%

Chemical Identifiers

4903-09-7
170.6
WYVGYYIZXPXHAZ-UHFFFAOYSA-N
21059
COC1=CC=C(C=O)C=C1Cl
C8H7ClO2
MFCD00016601
2-ethyl pyromeconic acid, 2-ethyl-3-hydroxy-4-pyrone, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethylpyromeconic acid, 3-hydroxy-2-ethyl-1,4-pyrone, 3-hydroxy-2-ethyl-4-pyrone, 4h-pyran-4-one, 2-ethyl-3-hydroxy, ethyl maltol, ethylmaltol, veltol plus
3-chloro-4-methoxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-532-2

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only