Learn More
3-chloro-4-methoxybenzaldehyde, 97%, ACROS Organics™
$261.74 - $261.74
Chemical Identifiers
CAS | 4903-09-7 |
---|---|
Molecular Formula | C8H7ClO2 |
Molecular Weight (g/mol) | 170.6 |
MDL Number | MFCD00016601 |
InChI Key | WYVGYYIZXPXHAZ-UHFFFAOYSA-N |
Synonym | 2-ethyl pyromeconic acid, 2-ethyl-3-hydroxy-4-pyrone, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethylpyromeconic acid, 3-hydroxy-2-ethyl-1,4-pyrone, 3-hydroxy-2-ethyl-4-pyrone, 4h-pyran-4-one, 2-ethyl-3-hydroxy, ethyl maltol, ethylmaltol, veltol plus |
PubChem CID | 21059 |
IUPAC Name | 3-chloro-4-methoxybenzaldehyde |
SMILES | COC1=CC=C(C=O)C=C1Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC435310010
|
Thermo Fisher Scientific
AC435310010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC435310050
|
Thermo Fisher Scientific
AC435310050 |
5g | Glass bottle |
Each for $261.74
|
|
Chemical Identifiers
4903-09-7 | |
170.6 | |
WYVGYYIZXPXHAZ-UHFFFAOYSA-N | |
21059 | |
COC1=CC=C(C=O)C=C1Cl |
C8H7ClO2 | |
MFCD00016601 | |
2-ethyl pyromeconic acid, 2-ethyl-3-hydroxy-4-pyrone, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethylpyromeconic acid, 3-hydroxy-2-ethyl-1,4-pyrone, 3-hydroxy-2-ethyl-4-pyrone, 4h-pyran-4-one, 2-ethyl-3-hydroxy, ethyl maltol, ethylmaltol, veltol plus | |
3-chloro-4-methoxybenzaldehyde |
Specifications
Authentic | |
128°C (5.0mmHg) | |
54°C to 56°C | |
4903-09-7 | |
100.0 | |
C8H7ClO2 | |
2-ethyl pyromeconic acid, 2-ethyl-3-hydroxy-4-pyrone, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethylpyromeconic acid, 3-hydroxy-2-ethyl-1,4-pyrone, 3-hydroxy-2-ethyl-4-pyrone, 4h-pyran-4-one, 2-ethyl-3-hydroxy, ethyl maltol, ethylmaltol, veltol plus | |
COC1=CC=C(C=O)C=C1Cl | |
170.6 | |
170.6 | |
97% |
Glass bottle | |
Brown to White | |
1g | |
96.0 | |
96% min. (GC) | |
MFCD00016601 | |
WYVGYYIZXPXHAZ-UHFFFAOYSA-N | |
3-chloro-4-methoxybenzaldehyde | |
21059 | |
Powder | |
3-chloro-4-methoxybenzaldehyde |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 225-532-2