Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-methoxybenzaldehyde, 96%

$70.82 - $244.09

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-methoxybenzaldehyde
CAS 4903-09-7
Molecular Formula C8H7ClO2
Formula Weight 170.6
MDL Number MFCD00016601
Synonym 3-Chloro-p-anisaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5044703

  SDS
alfa aesar™
H5044703
1g Each for $244.09

AAH50447MD

  SDS
alfa aesar™
H50447MD
250mg Each for $70.82
Description & Specifications

Specifications

Formula Weight 170.6
Chemical Name or Material 3-Chloro-4-methoxybenzaldehyde
Assay 96%
Molecular Formula C8H7ClO2
MDL Number MFCD00016601
CAS 4903-09-7
Synonym 3-Chloro-p-anisaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.