Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-4'-methoxyacetophenone, 96%

$83.43 - $264.76

Chemical Identifiers

Chemical Name or Material 3'-Chloro-4'-methoxyacetophenone
CAS 37612-52-5
Molecular Formula C9H9ClO2
Formula Weight 184.62
MDL Number MFCD04973413
Synonym 1-(3-Chloro-4-methoxyphenyl)ethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2582603

 
alfa aesar™
H2582603
1g Each for $83.43

AAH2582606

 
alfa aesar™
H2582606
5g Each for $264.76
Description & Specifications

Specifications

Formula Weight 184.62
Chemical Name or Material 3'-Chloro-4'-methoxyacetophenone
Assay 96%
Molecular Formula C9H9ClO2
MDL Number MFCD04973413
CAS 37612-52-5
Synonym 1-(3-Chloro-4-methoxyphenyl)ethanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 74° to 76°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.