missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-iodopyridine, 97%

Manufacturer:  Alfa Aesar™ H6386603

 View more versions of this product

Catalog No. AAH6386603



Specifications

Specifications

3-Chloro-4-iodopyridine
104°C to 108°C
77332-79-7
MFCD00272199
SQXBYRRNGPVSAU-UHFFFAOYSA-N
3-chloro-4-iodopyridine
12602200
Solid
Light Sensitive
1g
C5H3ClIN
pyridine, 3-chloro-4-iodo, pubchem9822, acmc-1birm, 3-chloro-4-iodo-pyridine, pyridine,3-chloro-4-iodo, ksc644e7r, 3-chloro-4-iodopyridine
C1=CN=CC(=C1I)Cl
239.44
239.44
97%

Chemical Identifiers

77332-79-7
239.44
SQXBYRRNGPVSAU-UHFFFAOYSA-N
12602200
C1=CN=CC(=C1I)Cl
C5H3ClIN
MFCD00272199
pyridine, 3-chloro-4-iodo, pubchem9822, acmc-1birm, 3-chloro-4-iodo-pyridine, pyridine,3-chloro-4-iodo, ksc644e7r, 3-chloro-4-iodopyridine
3-chloro-4-iodopyridine
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents