missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-iodobenzotrifluoride, 98%

$50.65 - $186.26

Chemical Identifiers

CAS 141738-80-9
Molecular Formula C7H3ClF3I
Formula Weight 306.45
MDL Number MFCD00042562
Synonym 2-Chloro-4-(trifluoromethyl)iodobenzene
Chemical Name or Material 3-Chloro-4-iodobenzotrifluoride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1468503 SDS Alfa Aesar™
A1468503
1g Each for $50.65
AAA1468506 SDS Alfa Aesar™
A1468506
5g Each for $186.26
Specifications Description & Specifications

Chemical Identifiers

141738-80-9
306.45
2-Chloro-4-(trifluoromethyl)iodobenzene
C7H3ClF3I
MFCD00042562
3-Chloro-4-iodobenzotrifluoride

Specifications

58° to 60°C (0.2mmHg)
141738-80-9
306.45
2-Chloro-4-(trifluoromethyl)iodobenzene
98%
1g
C7H3ClF3I
MFCD00042562
3-Chloro-4-iodobenzotrifluoride

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning