missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-iodobenzotrifluoride, 98%

Manufacturer:  Alfa Aesar™ A1468503

 View more versions of this product

Catalog No. AAA1468503



Specifications

Specifications

3-Chloro-4-iodobenzotrifluoride
1g
C7H3ClF3I
3-chloro-4-iodobenzotrifluoride, 2-chloro-1-iodo-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl iodobenzene, benzene, 2-chloro-1-iodo-4-trifluoromethyl, pubchem1973, acmc-209cms, buttpark 9857-21, 3-chloro-4-iodobenzotrifluoride, 2-chloro-4-trifluoromethyliodobenzene, 3-chloro-4-iodobenzotrifluoride
C1=CC(=C(C=C1C(F)(F)F)Cl)I
306.451
306.45
58°C to 60°C (0.2mmHg)
141738-80-9
MFCD00042562
ULJRBVGSKAKZKQ-UHFFFAOYSA-N
2-chloro-1-iodo-4-(trifluoromethyl)benzene
2736612
98%

Chemical Identifiers

141738-80-9
306.451
ULJRBVGSKAKZKQ-UHFFFAOYSA-N
2736612
C1=CC(=C(C=C1C(F)(F)F)Cl)I
C7H3ClF3I
MFCD00042562
3-chloro-4-iodobenzotrifluoride, 2-chloro-1-iodo-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl iodobenzene, benzene, 2-chloro-1-iodo-4-trifluoromethyl, pubchem1973, acmc-209cms, buttpark 9857-21, 3-chloro-4-iodobenzotrifluoride, 2-chloro-4-trifluoromethyliodobenzene, 3-chloro-4-iodobenzotrifluoride
2-chloro-1-iodo-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents