Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-iodoaniline, 98%

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-iodoaniline
CAS 135050-44-1
Molecular Formula C6H5ClIN
Formula Weight 253.47
MDL Number MFCD00051845
UN Number UN2811
Beilstein 3236464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1425606

 
alfa aesar™
A1425606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1425614

 
alfa aesar™
A1425614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1425622

 
alfa aesar™
A1425622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.47
Chemical Name or Material 3-Chloro-4-iodoaniline
Assay 98%
Molecular Formula C6H5ClIN
MDL Number MFCD00051845
CAS 135050-44-1
UN Number UN2811
Beilstein 3236464
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 65° to 71°C
Quantity 5g

Applications
3-Chloro-4-iodoaniline used as a intermediate for organic syntheses.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.