missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-hydroxypyridine, 98%

$20.82 - $266.78

Chemical Identifiers

CAS 89284-20-8
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD04114248
InChI Key NKEYLLFRPHHBOQ-UHFFFAOYSA-N
Synonym 3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
PubChem CID 2762929
IUPAC Name 3-chloro-1H-pyridin-4-one
SMILES C1=CNC=C(C1=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64783MD View Documents Alfa Aesar™
H64783MD
250mg Each for $20.82
 
AAH6478303 View Documents Alfa Aesar™
H6478303
1g Each for $61.05
 
AAH6478306 View Documents Alfa Aesar™
H6478306
5g Each for $266.78
 
Specifications

Chemical Identifiers

89284-20-8
129.543
NKEYLLFRPHHBOQ-UHFFFAOYSA-N
2762929
C1=CNC=C(C1=O)Cl
C5H4ClNO
MFCD04114248
3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
3-chloro-1H-pyridin-4-one

Specifications

204°C to 205°C
89284-20-8
MFCD04114248
3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
C1=CNC=C(C1=O)Cl
129.543
129.54
3-Chloro-4-hydroxypyridine
250mg
C5H4ClNO
1363515
NKEYLLFRPHHBOQ-UHFFFAOYSA-N
3-chloro-1H-pyridin-4-one
2762929
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning