missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-hydroxypyridine, 98%

Manufacturer:  Alfa Aesar™ H6478303

 View more versions of this product

Catalog No. AAH6478303



Specifications

Specifications

3-Chloro-4-hydroxypyridine
1g
C5H4ClNO
1363515
NKEYLLFRPHHBOQ-UHFFFAOYSA-N
3-chloro-1H-pyridin-4-one
2762929
98%
204°C to 205°C
89284-20-8
MFCD04114248
3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
C1=CNC=C(C1=O)Cl
129.543
129.54

Chemical Identifiers

89284-20-8
129.543
NKEYLLFRPHHBOQ-UHFFFAOYSA-N
2762929
C1=CNC=C(C1=O)Cl
C5H4ClNO
MFCD04114248
3-chloro-4-hydroxypyridine, 3-chloropyridin-4-ol, 4-pyridinol, 3-chloro, 3-chloropyridin-4 1h-one, 3-chloro-4-hydroxy pyridine, pubchem6680, 3-chloro-4-pyridinol, 3-chloropyridine-4-ol, acmc-20a0ep, 4 1h-pyridinone,3-chloro
3-chloro-1H-pyridin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents