Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-hydroxybenzoic acid, 97%, may contain up to ≈6% water

$142.24 - $184.72

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-hydroxybenzoic acid
CAS 3964-58-7
Molecular Formula C7H5ClO3
Formula Weight 172.57
MDL Number MFCD00002549
Beilstein 2207866
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1213414

  SDS
alfa aesar™
A1213414
25g Each for $142.24

AAA1213418

  SDS
alfa aesar™
A1213418
50g Each for $184.72
Description & Specifications

Specifications

Formula Weight 172.57
Chemical Name or Material 3-Chloro-4-hydroxybenzoic acid
Assay 97%
Molecular Formula C7H5ClO3
MDL Number MFCD00002549
CAS 3964-58-7
Beilstein 2207866
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration May contain up to ≈6% water
Melting Point 168° to 170°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.