missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-hydroxybenzoic acid, 97%, may contain up to ≈6% water

Manufacturer:  Alfa Aesar™ A1213418

 View more versions of this product

Catalog No. AAA1213418



Specifications

Specifications

3-Chloro-4-hydroxybenzoic acid
50g
C7H5ClO3
2207866
May contain up to ≈6% water
C1=CC(=C(C=C1C(=O)O)Cl)O
172.564
172.57
168°C to 170°C
3964-58-7
MFCD00002549
benzoic acid, 3-chloro-4-hydroxy, 3-chloro-4-hydroxy-benzoic acid, hemihydrate, pubchem13038, acmc-209vmh, 3-chloro-4hydroxybenzoic acid, 3-chloro4-hydroxybenzoic acid, 4-hydroxy-3-chlorobenzoic acid, 3-chloro-4-hydroxybenzoic scid, 3-chloro-4-hydroxy benzoic acid
QGNLHMKIGMZKJX-UHFFFAOYSA-N
3-chloro-4-hydroxybenzoic acid
19860
97%

Chemical Identifiers

3964-58-7
172.564
QGNLHMKIGMZKJX-UHFFFAOYSA-N
19860
C1=CC(=C(C=C1C(=O)O)Cl)O
C7H5ClO3
MFCD00002549
benzoic acid, 3-chloro-4-hydroxy, 3-chloro-4-hydroxy-benzoic acid, hemihydrate, pubchem13038, acmc-209vmh, 3-chloro-4hydroxybenzoic acid, 3-chloro4-hydroxybenzoic acid, 4-hydroxy-3-chlorobenzoic acid, 3-chloro-4-hydroxybenzoic scid, 3-chloro-4-hydroxy benzoic acid
3-chloro-4-hydroxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents