missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-hydroxyacetanilide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450080010

 View more versions of this product

Catalog No. AC450080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Chloro-4-hydroxyacetanilide
144.0°C to 146.0°C
3964-54-3
C8H8ClNO2
Solubility in water: soluble in hot water
CC(=O)NC1=CC(=C(C=C1)O)Cl
185.61
185.61
Glass Bottle
1g
97.5% min. (HPLC)
2-chloro-4-acetamidophenol, 3'-chloro-4'-hydroxyacetanilide, 3-chloro-4-hydroxyacet anilide, 3-chloro-4-hydroxyacetanilide, 4-acetylamino-2-chlorophenol, acetamide, n-3-chloro-4-hydroxyphenyl, chloro-4-acetamidophenol, n-3-chloro-4-hydroxyphenyl acetamide, paracetamol impurity c, zlchem 1169
JULKJDRBSRRBHT-UHFFFAOYSA-N
N-(3-chloro-4-hydroxyphenyl)acetamide
77579
98%

Chemical Identifiers

3964-54-3
185.61
2-chloro-4-acetamidophenol, 3'-chloro-4'-hydroxyacetanilide, 3-chloro-4-hydroxyacet anilide, 3-chloro-4-hydroxyacetanilide, 4-acetylamino-2-chlorophenol, acetamide, n-3-chloro-4-hydroxyphenyl, chloro-4-acetamidophenol, n-3-chloro-4-hydroxyphenyl acetamide, paracetamol impurity c, zlchem 1169
N-(3-chloro-4-hydroxyphenyl)acetamide
C8H8ClNO2
JULKJDRBSRRBHT-UHFFFAOYSA-N
77579
CC(=O)NC1=CC(=C(C=C1)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 223-570-4

SDS Documents