Please login to your online account to display your discounted pricing

3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C13H18BClO4
Formula Weight 284.55
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444680010

 
thermo fisher scientific
AC444680010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444680050

 
thermo fisher scientific
AC444680050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 284.55
Quantity 1g
Molecular Formula C13H18BClO4
Chemical Name or Material 3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester
Assay Percent Range 97%
CAS 1003298-84-7